Assay Details
Target Gene Details
Entrez Gene ID: | 12354 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
AV343731, Ca7 |
Location: |
Chr.8:104534700-104550347 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car7 | NM_001301164.1 | NP_001288093.1 | ||
NM_001301165.1 | NP_001288094.1 | |||
NM_053070.3 | NP_444300.1 | |||
AF291660.1 | AAG16230.1 | |||
AK090390.1 | ||||
AK144275.1 | BAE25808.1 | |||
AY075021.1 | AAL78169.1 | |||
BC094913.1 | AAH94913.1 |