Assay Details
Target Gene Details
Entrez Gene ID: | 66498 |
Gene Name: | DET1 and DDB1 associated 1 |
Gene Aliases: |
1500034J01Rik, 1700095J19Rik, R74921 |
Location: |
Chr.8:71469171-71476097 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dda1 | NM_001294258.1 | NP_001281187.1 | ||
NM_001294259.1 | NP_001281188.1 | |||
NM_025600.2 | NP_079876.1 | |||
NR_122114.1 | ||||
NR_122115.1 | ||||
AA036394.1 | ||||
AK006318.1 | BAB24524.1 | |||
AK007085.1 | ||||
AK032966.1 | BAC28103.1 | |||
AK149117.1 | ||||
AK149980.1 | BAE29210.1 | |||
AK160132.1 | ||||
AK168085.1 | BAE40058.1 | |||
AK186341.1 | ||||
BB610194.1 | ||||
BC034701.1 | AAH34701.1 | |||
BC093523.1 | AAH93523.1 | |||
BY297077.1 | ||||
BY316490.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |