Assay Details
Target Gene Details
Entrez Gene ID: | 102032 |
Gene Name: | small integral membrane protein 19 |
Gene Aliases: |
AI316807, BG694986, EST-c39 |
Location: |
Chr.8:22462614-22476879 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smim19 | NM_001012667.2 | NP_001012685.2 | ||
NM_001146117.1 | NP_001139589.1 | |||
AK131902.1 | BAE20860.1 | |||
AK133712.1 | BAE21793.1 | |||
AK140957.1 | BAE24529.1 | |||
AK178820.1 | ||||
AK213440.1 | ||||
AK218393.1 | ||||
BB618223.1 | ||||
BC048089.1 | AAH48089.1 | |||
BQ922100.1 |