Assay Details
Target Gene Details
Entrez Gene ID: | 353188 |
Gene Name: | a disintegrin and metallopeptidase domain 32 |
Gene Aliases: |
- |
Location: |
Chr.8:24836143-24948917 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adam32 | NM_001293693.1 | 6 | 404 | NP_001280622.1 |
NM_153397.2 | 6 | 404 | NP_700446.2 | |
AF513715.1 | 6 | 387 | AAN03858.1 | |
AK005759.1 | 6 | 395 | BAC25124.1 | |
BC060983.1 | 6 | 401 | AAH60983.1 |
More Information
Set Membership: |
Intragenic Exonic |