Assay Details
Target Gene Details
Entrez Gene ID: | 353188 |
Gene Name: | a disintegrin and metallopeptidase domain 32 |
Gene Aliases: |
- |
Location: |
Chr.8:24836143-24948917 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adam32 | NM_001293693.1 | NP_001280622.1 | ||
NM_153397.2 | NP_700446.2 | |||
AF513715.1 | AAN03858.1 | |||
AK005759.1 | BAC25124.1 | |||
BC060983.1 | AAH60983.1 | |||
KF144343.1 | ||||
KF144345.1 |
More Information
Set Membership: |
Intragenic Non-exonic |