Assay Details
Target Gene Details
Entrez Gene ID: | 234421 |
Gene Name: | calcium and integrin binding family member 3 |
Gene Aliases: |
C730014M21Rik, Gm1107, KIP3 |
Location: |
Chr.8:72192699-72213092 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cib3 | NM_001080812.1 | 2 | 155 | NP_001074281.1 |
AK050094.1 | 2 | 155 | ||
BB870365.1 | 2 | 180 | ||
BC120812.1 | 2 | 137 | AAI20813.1 | |
BC120814.1 | 2 | 137 | AAI20815.1 |