Assay Details
Target Gene Details
Entrez Gene ID: | 170780 |
Gene Name: | CD209e antigen |
Gene Aliases: |
SIGNR4, mSIGNR4 |
Location: |
Chr.8:3847973-3854286 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209e | NM_130905.2 | 4 | 319 | NP_570975.1 |
AF373412.1 | 4 | 319 | AAL13238.1 | |
AK040917.1 | 4 | 341 | BAC30744.1 | |
BC107188.2 | 4 | 319 | AAI07189.1 | |
BC107189.2 | 4 | 319 | AAI07190.1 |
More Information
Set Membership: |
Intragenic Exonic |