Assay Details
Target Gene Details
Entrez Gene ID: | 20924 |
Gene Name: | suppressor of Ty 5 |
Gene Aliases: |
AL033283, AL033321, AU019549, Spt5, Supt5h |
Location: |
Chr.7:28314891-28338804 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supt5 | NM_013676.1 | 14 | 1115 | NP_038704.1 |
AK145117.1 | 14 | 1155 | BAE26244.1 | |
AK145465.1 | ||||
AK146055.1 | 14 | 1143 | BAE26864.1 | |
AK146453.1 | 14 | 1143 | BAE27184.1 | |
AK146583.1 | 13 | 1362 | BAE27278.1 | |
AK146650.1 | 14 | 1156 | BAE27330.1 | |
AK159786.1 | 15 | 1240 | ||
AK177285.1 | 3 | 144 | ||
AK186836.1 | 2 | 88 | ||
BC007132.1 | 14 | 1115 | AAH07132.1 | |
BC057449.1 | 14 | 1130 | AAH57449.1 | |
BC058598.1 | 13 | 1090 | AAH58598.1 | |
BC059849.1 | 6 | 698 | AAH59849.1 | |
U88539.1 | 14 | 1142 | AAC40052.1 |