Assay Details
Target Gene Details
Entrez Gene ID: | 22282 |
Gene Name: | upstream transcription factor 2 |
Gene Aliases: |
Usf-2, bHLHb12 |
Location: |
Chr.7:30945248-30957250 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usf2 | NM_011680.2 | 3 | 217 | NP_035810.1 |
AB221562.1 | 3 | 176 | ||
AK146871.1 | 3 | 164 | BAE27494.1 | |
AK158906.1 | ||||
BC082995.1 | 3 | 217 | AAH82995.1 | |
U01662.1 | 3 | 233 | AAA20492.1 | |
U01663.1 | AAA20493.1 |