Assay Details
Target Gene Details
Entrez Gene ID: | 19943 |
Gene Name: | ribosomal protein L28 |
Gene Aliases: |
D7Wsu21e |
Location: |
Chr.7:4792888-4794562 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpl28 | NM_009081.2 | NP_033107.1 | ||
AK012368.1 | BAB28192.1 | |||
AK018714.1 | BAB31362.1 | |||
AK076133.1 | 5 | 516 | BAC36209.1 | |
BC024395.1 | AAH24395.1 | |||
BC086797.1 | AAH86797.1 | |||
BC086932.1 | AAH86932.1 | |||
BC092012.1 | AAH92012.1 | |||
X74856.1 | CAA52848.1 |
More Information
Set Membership: |
Intragenic Non-exonic |