Assay Details
Target Gene Details
Entrez Gene ID: | 75613 |
Gene Name: | mediator complex subunit 25 |
Gene Aliases: |
2610034E13Rik, 2610529E18Rik, ESTM2 |
Location: |
Chr.7:44876765-44892664 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med25 | NM_029365.2 | NP_083641.1 | ||
AK011662.1 | ||||
AK089940.1 | ||||
AK090026.1 | ||||
AK158238.1 | ||||
AK165760.1 | ||||
AK182054.1 | ||||
BC021333.1 | AAH21333.1 | |||
BC031138.1 | AAH31138.1 | |||
BC071206.1 | AAH71206.1 |
More Information
Set Membership: |
Intragenic Non-exonic |