Assay Details
Target Gene Details
Entrez Gene ID: | 269951 |
Gene Name: | isocitrate dehydrogenase 2 (NADP+), mitochondrial |
Gene Aliases: |
E430004F23, IDPm, Idh-2 |
Location: |
Chr.7:80094846-80115387 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idh2 | NM_173011.2 | 9 | 1196 | NP_766599.2 |
AF212319.1 | 9 | 1167 | AAG43538.1 | |
AK088110.1 | 9 | 1168 | BAC40149.1 | |
AK145753.1 | 9 | 1141 | BAE26628.1 | |
AK146564.1 | 9 | 1282 | ||
AK161640.1 | 9 | 1180 | BAE36505.1 | |
AK169395.1 | 9 | 1193 | BAE41142.1 | |
BC060030.1 | 9 | 1184 | AAH60030.1 |
More Information
Set Membership: |
Intragenic Exonic |