Assay Details
Target Gene Details
Entrez Gene ID: | 110826 |
Gene Name: | electron transferring flavoprotein, beta polypeptide |
Gene Aliases: |
0610009I16Rik, 2810441H06Rik |
Location: |
Chr.7:43444072-43457800 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Etfb | NM_026695.3 | NP_080971.2 | ||
NR_075104.1 | ||||
AA207939.1 | ||||
AK002407.1 | BAB22076.1 | |||
AK013284.1 | ||||
AK150293.1 | BAE29447.1 | |||
BC049237.1 | AAH49237.1 | |||
BC069877.1 | AAH69877.1 |
More Information
Set Membership: |
Intragenic Non-exonic |