Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcgrt | NM_010189.3 | 1 | 100 | NP_034319.2 |
AK008167.1 | 1 | 74 | ||
AK008360.1 | 1 | 74 | ||
AK146933.1 | 1 | 61 | ||
AK152879.1 | BAE31564.1 | |||
BC003786.1 | AAH03786.1 | |||
BY063885.1 | 1 | 131 | ||
D37874.1 | BAA07111.1 |
More Information
Set Membership: |
Intragenic Exonic |