Assay Details
Target Gene Details
Entrez Gene ID: | 57442 |
Gene Name: | potassium voltage-gated channel, Isk-related subfamily, gene 3 |
Gene Aliases: |
2210017H05Rik, MiRP2 |
Location: |
Chr.7:100176507-100184869 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcne3 | NM_001190869.1 | NP_001177798.1 | ||
NM_001190870.1 | NP_001177799.1 | |||
NM_001190871.2 | NP_001177800.1 | |||
NM_001190950.1 | NP_001177879.1 | |||
NM_020574.5 | NP_065599.1 | |||
AF076532.1 | AAD28090.1 | |||
AK008744.1 | BAB25871.1 | |||
AK132773.1 | BAE21350.1 | |||
BC004629.1 | AAH04629.1 | |||
BX514261.1 | ||||
DQ832280.1 | ||||
DV056972.1 | ||||
EH094997.1 | ||||
EH101376.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)