Assay Details
Target Gene Details
Entrez Gene ID: | 16202 |
Gene Name: | integrin linked kinase |
Gene Aliases: |
AA511515, ESTM24 |
Location: |
Chr.7:105736590-105742925 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilk | NM_001161724.1 | NP_001155196.1 | ||
NM_010562.2 | NP_034692.2 | |||
AK002344.1 | ||||
AK011766.1 | ||||
AK154207.1 | ||||
AK216132.1 | ||||
BC003737.1 | AAH03737.1 | |||
U94479.1 | AAB94646.1 |
More Information
Set Membership: |
Intragenic Non-exonic |