Assay Details
Target Gene Details
Entrez Gene ID: | 56209 |
Gene Name: | glycerophosphodiester phosphodiesterase 1 |
Gene Aliases: |
1200003M13Rik, Mir16, RGS16 |
Location: |
Chr.7:118688558-118705738 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gde1 | NM_019580.4 | 5 | 789 | NP_062526.1 |
AF212860.1 | 5 | 770 | AAF65232.1 | |
AK004583.1 | 5 | 802 | ||
AK005361.1 | 5 | 790 | BAB23975.1 | |
AK150807.1 | 5 | 811 | BAE29870.1 | |
AK187418.1 | 2 | 82 | ||
AK198453.1 | 2 | 82 | ||
AK205654.1 | 2 | 82 | ||
BC003902.1 | 5 | 748 | AAH03902.1 |
More Information
Set Membership: |
Intragenic Exonic |