Assay Details
Target Gene Details
Entrez Gene ID: | 56209 |
Gene Name: | glycerophosphodiester phosphodiesterase 1 |
Gene Aliases: |
1200003M13Rik, Mir16, RGS16 |
Location: |
Chr.7:118688558-118705738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gde1 | NM_019580.4 | NP_062526.1 | ||
AF212860.1 | AAF65232.1 | |||
AK004583.1 | ||||
AK005361.1 | BAB23975.1 | |||
AK150807.1 | BAE29870.1 | |||
AK187418.1 | ||||
AK198453.1 | ||||
AK205654.1 | ||||
BC003902.1 | AAH03902.1 |
More Information
Set Membership: |
Intragenic Non-exonic |