Assay Details
Target Gene Details
Entrez Gene ID: | 19144 |
Gene Name: | kallikrein related-peptidase 6 |
Gene Aliases: |
AI849898, BSP, Bssp, Klk29, MSP, Prss18, Prss9, neurosin |
Location: |
Chr.7:43824544-43832027 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klk6 | NM_001164696.1 | 4 | 343 | NP_001158168.1 |
NM_001164697.1 | 4 | 339 | NP_001158169.1 | |
NM_001164698.1 | 4 | 240 | NP_001158170.1 | |
NM_011177.2 | 3 | 351 | NP_035307.2 | |
AB008928.1 | 4 | 329 | BAB55605.1 | |
AB015206.1 | 4 | 327 | BAA28895.1 | |
AK138218.1 | 4 | 340 | BAE23586.1 | |
BC031119.1 | 3 | 188 | AAH31119.1 | |
EL956218.1 | 4 | 215 | ||
Y18723.1 | 3 | 351 | CAA77269.1 |
More Information
Set Membership: |
Intragenic Exonic |