Assay Details
Target Gene Details
Entrez Gene ID: | 233876 |
Gene Name: | HIRA interacting protein 3 |
Gene Aliases: |
B130036O03, C86302 |
Location: |
Chr.7:126861972-126865377 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hirip3 | NM_172746.3 | NP_766334.1 | ||
AK045122.1 | BAC32232.1 | |||
AK135526.1 | ||||
AK161657.1 | ||||
AK162096.1 | BAE36722.1 | |||
BC055687.1 | AAH55687.1 |