Assay Details
Target Gene Details
Entrez Gene ID: | 333182 |
Gene Name: | cytochrome c oxidase subunit VIb polypeptide 2 |
Gene Aliases: |
1700067P11Rik, BC048670, COXVIB2 |
Location: |
Chr.7:4751792-4753094 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox6b2 | NM_001289848.1 | NP_001276777.1 | ||
NM_001289849.1 | NP_001276778.1 | |||
NM_001289850.1 | NP_001276779.1 | |||
NM_183405.2 | NP_899664.1 | |||
NM_183406.3 | NP_899665.1 | |||
NR_110411.1 | ||||
AK160152.1 | BAE35659.1 | |||
AW123567.1 | ||||
AY152400.1 | AAN46753.1 | |||
BC048670.1 | AAH48670.1 | |||
BC107296.1 | AAI07297.1 | |||
BC107297.1 | AAI07298.1 | |||
BC125562.1 | ||||
BC125564.1 | ||||
BC145201.1 | ||||
BG080371.2 |
More Information
Set Membership: |
Intragenic Non-exonic |