Assay Details
Target Gene Details
Entrez Gene ID: | 333182 |
Gene Name: | cytochrome c oxidase subunit VIb polypeptide 2 |
Gene Aliases: |
1700067P11Rik, BC048670, COXVIB2 |
Location: |
Chr.7:4751792-4753094 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox6b2 | NM_001289848.1 | 2 | 254 | NP_001276777.1 |
NM_001289849.1 | 2 | 223 | NP_001276778.1 | |
NM_001289850.1 | 2 | 223 | NP_001276779.1 | |
NM_183405.2 | 2 | 223 | NP_899664.1 | |
NM_183406.3 | 1 | 134 | NP_899665.1 | |
NR_110411.1 | 2 | 223 | ||
AK160152.1 | 2 | 153 | BAE35659.1 | |
AW123567.1 | 3 | 299 | ||
AY152400.1 | 2 | 223 | AAN46753.1 | |
BC048670.1 | 1 | 134 | AAH48670.1 | |
BC107296.1 | 2 | 180 | AAI07297.1 | |
BC107297.1 | 2 | 180 | AAI07298.1 | |
BC125562.1 | 2 | 163 | ||
BC125564.1 | 2 | 166 | ||
BC145201.1 | 2 | 198 | ||
BG080371.2 | 2 | 131 | ||
CT010366.1 | 1 | 67 | CAJ18573.1 | |
W88135.1 | 1 | 210 |
More Information
Set Membership: |
Intragenic Exonic |