Assay Details
Target Gene Details
Entrez Gene ID: | 69170 |
Gene Name: | RIKEN cDNA 1810026B05 gene |
Gene Aliases: |
- |
Location: |
Chr.7:73539798-73558395 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1810026B05Rik | NR_037569.1 | |||
AK007604.1 | ||||
AK140938.1 |