Assay Details
Target Gene Details
Entrez Gene ID: | 20901 |
Gene Name: | serine/threonine kinase receptor associated protein |
Gene Aliases: |
AW557906, C78091, C79202, Unrip |
Location: |
Chr.6:137735082-137751930 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Strap | NM_011499.3 | NP_035629.2 | ||
AF096285.1 | AAC98300.1 | |||
AK075804.1 | BAC35972.1 | |||
AK077504.1 | BAC36834.1 | |||
AK145971.1 | BAE26796.1 | |||
AK147179.1 | ||||
AK152283.1 | BAE31097.1 | |||
AK154158.1 | ||||
AK210373.1 | ||||
BC141076.1 | ||||
BC145342.1 |
More Information
Set Membership: |
Intragenic Non-exonic |