Assay Details
Target Gene Details
Entrez Gene ID: | 67442 |
Gene Name: | retinol saturase (all trans retinol 13,14 reductase) |
Gene Aliases: |
0610039N19Rik, C80029, MMT-7, Ppsig |
Location: |
Chr.6:72598628-72607488 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Retsat | NM_026159.4 | 5 | 895 | NP_080435.3 |
AK002851.1 | 2 | 247 | BAB22406.2 | |
AY704159.1 | 5 | 895 | AAU25836.1 | |
BC011203.1 | 1 | 77 | ||
BC117750.1 | 5 | 890 | ||
BC117751.1 | 5 | 895 | ||
EF620365.1 | 5 | 921 |