Assay Details
Target Gene Details
Entrez Gene ID: | 24110 |
Gene Name: | ubiquitin specific peptidase 18 |
Gene Aliases: |
1110058H21Rik, AW047653, UBP43, Ubp15 |
Location: |
Chr.6:121245906-121270917 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp18 | NM_011909.2 | NP_036039.2 | ||
AF069502.1 | AAD21222.1 | |||
AK004296.1 | ||||
AK149772.1 | BAE29076.1 | |||
AK151011.1 | BAE30032.1 | |||
AK151260.1 | BAE30248.1 | |||
AK151722.1 | BAE30640.1 | |||
AK152034.1 | BAE30893.1 | |||
AK152574.1 | BAE31326.1 | |||
AK153312.1 | BAE31893.1 | |||
BC138562.1 | ||||
BC138577.1 | ||||
CT010228.1 | CAJ18436.1 |
More Information
Set Membership: |
Intragenic Non-exonic |