Assay Details
Target Gene Details
Entrez Gene ID: | 21333 |
Gene Name: | tachykinin 1 |
Gene Aliases: |
4930528L02Rik, NK-1, NK1, Nkna, PPT-A, PPTA, SP |
Location: |
Chr.6:7554776-7562978 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tac1 | NM_009311.2 | NP_033337.1 | ||
BC117081.1 | ||||
BC119122.1 | ||||
BC144737.1 | ||||
D17584.1 | BAA04508.1 | |||
M68908.1 | AAA39969.1 | |||
M68909.1 | AAA39970.1 |