Assay Details
Target Gene Details
Entrez Gene ID: | 18575 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
- |
Location: |
Chr.6:56069798-56652516 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde1c | NM_001025568.2 | NP_001020739.1 | ||
NM_001159952.1 | NP_001153424.1 | |||
NM_001159953.1 | NP_001153425.1 | |||
NM_001159955.1 | NP_001153427.1 | |||
NM_001159956.1 | NP_001153428.1 | |||
NM_001159957.1 | NP_001153429.1 | |||
NM_001159960.1 | NP_001153432.1 | |||
NM_011054.4 | NP_035184.1 | |||
AK014887.1 | BAB29604.1 | |||
AK015375.1 | ||||
AK029531.1 | BAC26499.1 | |||
AK030423.1 | BAC26956.1 | |||
AK050499.1 | ||||
AK077123.1 | BAC36626.1 | |||
AK082180.1 | BAC38431.1 | |||
AK082497.1 | ||||
AK133243.1 | ||||
AK139923.1 | BAE24184.1 | |||
BC032277.1 | AAH32277.1 | |||
L76944.1 | AAC37701.1 | |||
L76946.1 | AAC37703.1 | |||
L76947.1 | AAC37702.1 |
More Information
Set Membership: |
Intragenic Non-exonic |