Assay Details
Target Gene Details
Entrez Gene ID: | 57377 |
Gene Name: | mannosyl-oligosaccharide glucosidase |
Gene Aliases: |
1810017N02Rik, AI181835, Gcs1 |
Location: |
Chr.6:83115506-83118902 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mogs | NM_020619.2 | NP_065644.2 | ||
AK144946.1 | BAE26149.1 | |||
AK157286.1 | BAE34029.1 | |||
AK219249.1 | ||||
BC051949.1 | AAH51949.1 |
More Information
Set Membership: |
Intragenic Non-exonic |