Assay Details
Target Gene Details
Entrez Gene ID: | 53621 |
Gene Name: | CCR4-NOT transcription complex, subunit 4 |
Gene Aliases: |
Not4, Not4h, Not4hp |
Location: |
Chr.6:35022065-35133737 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnot4 | NM_001164411.1 | NP_001157883.1 | ||
NM_001164412.1 | NP_001157884.1 | |||
NM_001164413.1 | NP_001157885.1 | |||
NM_001164414.1 | NP_001157886.1 | |||
NM_016877.4 | NP_058573.3 | |||
AK028190.1 | BAC25801.1 | |||
AK029753.1 | ||||
AK032248.1 | BAC27779.1 | |||
AK075887.1 | ||||
AK144760.1 | ||||
AK165997.1 | BAE38508.1 | |||
BC048243.1 | ||||
BC058778.1 | AAH58778.1 | |||
U71269.1 | AAD00181.1 |
More Information
Set Membership: |
Intragenic Non-exonic |