Assay Details
Target Gene Details
Entrez Gene ID: | 78910 |
Gene Name: | ankyrin repeat and SOCS box-containing 15 |
Gene Aliases: |
4930400E23Rik, Asb-15 |
Location: |
Chr.6:24528144-24573164 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asb15 | NM_080847.3 | 6 | 659 | NP_543123.2 |
AF403043.1 | 6 | 660 | AAL57362.1 | |
AK019556.1 | 6 | 660 | ||
AK029759.1 | 6 | 591 | BAC26602.1 | |
AK079418.1 | 6 | 765 | BAC37641.1 | |
BC050794.1 | 6 | 608 | AAH50794.1 |