Assay Details
Target Gene Details
Entrez Gene ID: | 78910 |
Gene Name: | ankyrin repeat and SOCS box-containing 15 |
Gene Aliases: |
4930400E23Rik, Asb-15 |
Location: |
Chr.6:24528144-24573164 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asb15 | NM_080847.3 | NP_543123.2 | ||
AF403043.1 | AAL57362.1 | |||
AK019556.1 | ||||
AK029759.1 | BAC26602.1 | |||
AK079418.1 | BAC37641.1 | |||
BC050794.1 | AAH50794.1 |