Assay Details
Target Gene Details
Entrez Gene ID: | 77574 |
Gene Name: | TRPM8 channel-associated factor 1 |
Gene Aliases: |
2810407D09Rik, 3321401G04Rik, A230020K05Rik, AI848529, Fam115a |
Location: |
Chr.6:42667999-42710110 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcaf1 | NM_029930.2 | 5 | 1966 | NP_084206.1 |
AK014379.1 | 5 | 1495 | BAC25433.1 | |
AK053509.1 | 5 | 1967 | BAC35410.1 | |
AK076242.1 | 5 | 1912 | ||
AK083917.1 | 5 | 1914 | BAC39061.1 | |
AK132148.1 | 5 | 1934 | BAE20997.1 | |
AK148593.1 | 5 | 1945 | ||
AK173013.1 | 5 | 1948 | BAD32291.1 | |
BC031718.1 | 5 | 1994 | AAH31718.1 | |
BC043304.1 | 3 | 226 | AAH43304.1 |