Assay Details
Target Gene Details
Entrez Gene ID: | 75788 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 1 |
Gene Aliases: |
4930431E10Rik, mKIAA1625 |
Location: |
Chr.5:144876495-144965830 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smurf1 | NM_001038627.1 | 14 | 1894 | NP_001033716.1 |
NM_029438.3 | 14 | 1894 | NP_083714.3 | |
AK015264.1 | 10 | 1293 | BAB29770.2 | |
AK034736.1 | 5 | 492 | BAC28813.1 | |
AK122534.1 | 3 | 269 | BAC65816.1 | |
AK154491.1 | 14 | 1895 | BAE32623.1 | |
BC029097.1 | 14 | 1922 | AAH29097.1 |
More Information
Set Membership: |
Intragenic Exonic |