Assay Details
Target Gene Details
Entrez Gene ID: | 20817 |
Gene Name: | serine/arginine-rich protein specific kinase 2 |
Gene Aliases: |
AW226533, AW492537, AW547358, Wbp6, mSRPK2 |
Location: |
Chr.5:23503356-23684641 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srpk2 | NM_009274.2 | 4 | 551 | NP_033300.2 |
AB006036.1 | 4 | 344 | BAA24055.1 | |
AK014004.1 | 5 | 470 | ||
AK047059.1 | 1 | 2348 | ||
AK136861.1 | 4 | 516 | ||
AK161384.1 | 5 | 610 | BAE36363.1 | |
AK169580.1 | 4 | 524 | BAE41239.1 | |
BC020178.1 | 4 | 448 | AAH20178.1 | |
BC062941.1 | 5 | 496 | AAH62941.1 | |
U92456.1 | 5 | 473 | AAC53193.1 |
More Information
Set Membership: |
Intragenic Exonic |