Assay Details
Target Gene Details
Entrez Gene ID: | 100763 |
Gene Name: | ubiquitin protein ligase E3C |
Gene Aliases: |
AI853514, mKIAA0010 |
Location: |
Chr.5:29569242-29676077 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube3c | NM_133907.3 | 18 | 2675 | NP_598668.1 |
AK029973.1 | 18 | 2485 | BAC26709.1 | |
AK046056.1 | 18 | 2676 | BAC32585.1 | |
AK049125.1 | 1 | 66 | BAC33557.1 | |
AK122187.1 | 18 | 2610 | BAC65469.1 | |
AK185151.1 | ||||
AK203997.1 | 1 | 146 | ||
BC021525.1 | 7 | 920 | AAH21525.1 | |
BC120731.1 | 18 | 2422 | ||
BC137626.1 | 18 | 2422 |
More Information
Set Membership: |
Intragenic Exonic |