Assay Details
Target Gene Details
Entrez Gene ID: | 76571 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
1700011C14Rik, Dusp24, Mkstyx |
Location: |
Chr.5:135747220-135778410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Styxl1 | NM_001289554.1 | NP_001276483.1 | ||
NM_001289555.1 | NP_001276484.1 | |||
NM_001289556.1 | NP_001276485.1 | |||
NM_001289557.1 | NP_001276486.1 | |||
NM_029659.4 | NP_083935.3 | |||
AK005599.1 | BAB24144.1 | |||
AK005858.1 | BAB24279.1 | |||
AK161310.1 | BAE36314.1 | |||
BC045148.1 | AAH45148.1 | |||
BF471066.1 |