Assay Details
Target Gene Details
Entrez Gene ID: | 22697 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
AI326272, CTfin51, RU49, Zfp-38, Zfp38, Zipro1 |
Location: |
Chr.5:138116899-138134265 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zscan21 | NM_001044703.2 | 3 | 873 | NP_001038168.1 |
NM_001044704.2 | 4 | 939 | NP_001038169.1 | |
NM_001044705.2 | 3 | 854 | NP_001038170.1 | |
NM_011757.3 | 4 | 958 | NP_035887.2 | |
AK146776.1 | 4 | 932 | ||
AK146875.1 | 4 | 919 | BAE27498.1 | |
BC010591.1 | 3 | 801 | AAH10591.1 | |
BU513148.1 | 3 | 815 | ||
D10630.1 | 3 | 792 | BAA01480.1 | |
U41671.1 | 4 | 889 | AAB03786.1 | |
X63747.1 | 3 | 1649 | CAA45280.1 |
More Information
Set Membership: |
Intragenic Exonic |