Assay Details
Target Gene Details
Entrez Gene ID: | 20349 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E |
Gene Aliases: |
6430702L12, AA408817, Semah, mKIAA0331 |
Location: |
Chr.5:14025276-14256689 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema3e | NM_011348.2 | 10 | 1706 | NP_035478.2 |
AF034744.1 | 10 | 1441 | AAD01996.1 | |
AK031300.1 | 1 | 76 | ||
AK129116.1 | 5 | 471 | BAC97926.1 | |
AK141295.1 | 9 | 2428 | ||
AK142340.1 | 10 | 1533 | BAE25038.1 | |
AK159214.1 | 10 | 1497 | BAE34903.1 | |
BC057956.1 | 10 | 1604 | AAH57956.1 | |
Z80941.1 | 10 | 1072 | CAB02590.1 | |
Z93947.1 | 10 | 1681 | CAB07987.1 | |
Z93948.1 | 10 | 1681 | CAB07988.1 |
More Information
Set Membership: |
Intragenic Exonic |