Assay Details
Target Gene Details
Entrez Gene ID: | 320951 |
Gene Name: | phosphatidylserine decarboxylase |
Gene Aliases: |
9030221M09Rik |
Location: |
Chr.5:32736303-32785636 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pisd | NM_177298.3 | NP_796272.2 | ||
AK033483.1 | BAC28313.1 | |||
AK034656.1 | BAC28786.1 | |||
AK162028.1 | BAE36689.1 | |||
AK162737.1 | BAE37044.1 | |||
AK167554.1 | BAE39619.1 | |||
BC070408.1 | AAH70408.1 | |||
BC094594.1 | AAH94594.1 | |||
BC116695.1 | ||||
BC117509.1 | ||||
CX201846.1 |
More Information
Set Membership: |
Intragenic Non-exonic |