Assay Details
Target Gene Details
Entrez Gene ID: | 18647 |
Gene Name: | cyclin-dependent kinase 14 |
Gene Aliases: |
Pftk1, mKIAA0834 |
Location: |
Chr.5:4803384-5380251 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk14 | NM_011074.2 | NP_035204.2 | ||
AF033655.1 | AAB87504.1 | |||
AK016370.1 | BAB30208.1 | |||
AK045083.1 | BAC32212.1 | |||
AK051283.1 | BAE43346.1 | |||
AK051411.1 | BAC34630.1 | |||
AK083790.1 | BAC39020.1 | |||
AK129226.1 | BAC98036.1 |
More Information
Set Membership: |
Intragenic Non-exonic |