Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkag2 | NM_001170555.1 | 8 | 791 | NP_001164026.1 |
NM_001170556.1 | 8 | 788 | NP_001164027.1 | |
NM_145401.2 | 12 | 1870 | NP_663376.2 | |
AK032238.1 | 10 | 1382 | BAC27775.1 | |
AK034765.1 | 6 | 456 | BAC28827.1 | |
AK154296.1 | 12 | 1871 | BAE32497.1 | |
AK165834.1 | 8 | 730 | BAE38403.1 | |
AY348864.1 | 8 | 721 | AAQ55224.1 | |
BC015283.1 | 12 | 1658 | AAH15283.1 | |
BQ572978.1 | 8 | 721 | ||
BU707674.1 | 8 | 691 |
More Information
Set Membership: |
Intragenic Exonic |