Assay Details
Target Gene Details
Entrez Gene ID: | 14886 |
Gene Name: | general transcription factor II I |
Gene Aliases: |
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6 |
Location: |
Chr.5:134237832-134314760 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2i | NM_001080746.2 | 21 | 2188 | NP_001074215.1 |
NM_001080747.2 | 20 | 2125 | NP_001074216.1 | |
NM_001080748.2 | 19 | 2068 | NP_001074217.1 | |
NM_001080749.2 | 19 | 2068 | NP_001074218.1 | |
NM_010365.4 | 20 | 2131 | NP_034495.2 | |
AF017085.1 | 21 | 1832 | AAC53569.1 | |
AF043219.1 | 19 | 1752 | AAC02990.1 | |
AF043220.1 | 18 | 1689 | AAC02991.1 | |
AK145698.1 | 11 | 1016 | ||
AK147201.1 | 19 | 2026 | BAE27759.1 | |
AY030290.1 | 20 | 1818 | AAK49785.1 | |
AY030291.1 | 20 | 1824 | AAK49786.1 | |
AY030292.1 | 19 | 1761 | AAK49787.1 | |
AY030293.1 | 21 | 1881 | AAK49788.1 | |
BC053044.1 | 21 | 2157 | AAH53044.1 |
More Information
Set Membership: |
Intragenic Exonic |