Assay Details
Target Gene Details
Entrez Gene ID: | 15932 |
Gene Name: | iduronidase, alpha-L- |
Gene Aliases: |
6030426D08 |
Location: |
Chr.5:108660331-108685446 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idua | NM_008325.4 | NP_032351.2 | ||
NM_172997.1 | NP_766585.1 | |||
AK031414.1 | BAC27393.1 | |||
AK045330.1 | BAC32313.1 | |||
AK155627.1 | BAE33353.1 | |||
AK169998.1 | BAE41504.1 | |||
BC057935.1 | AAH57935.1 | |||
L34111.1 | AAC42044.1 |
Target Gene Details
Entrez Gene ID: | 231583 |
Gene Name: | solute carrier family 26 (sulfate transporter), member 1 |
Gene Aliases: |
Sat1 |
Location: |
Chr.5:108669970-108678097 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc26a1 | AY093420.1 | 3 | 3424 | AAM18183.1 |
More Information
Set Membership: |
Intragenic Non-exonic |