Assay Details
Target Gene Details
Entrez Gene ID: | 15234 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF |
Location: |
Chr.5:16553335-16622408 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hgf | NM_001289458.1 | NP_001276387.1 | ||
NM_001289459.1 | NP_001276388.1 | |||
NM_001289460.1 | NP_001276389.1 | |||
NM_001289461.1 | NP_001276390.1 | |||
NM_010427.5 | NP_034557.3 | |||
AF042856.1 | AAC40051.1 | |||
AK042121.1 | BAC31175.1 | |||
AK052289.1 | ||||
AK082461.1 | BAC38497.1 | |||
AK142159.1 | ||||
BC119228.1 | ||||
D10212.1 | BAA01064.1 | |||
D10213.1 | BAA01065.1 | |||
S71816.1 | AAB31855.1 | |||
X72307.1 | CAA51054.1 | |||
X84046.1 | CAA58865.1 |
More Information
Set Membership: |
Intragenic Non-exonic |