Assay Details
Target Gene Details
Entrez Gene ID: | 381633 |
Gene Name: | predicted gene 1673 |
Gene Aliases: |
- |
Location: |
Chr.5:33983472-33985009 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm1673 | NM_001033458.3 | NP_001028630.1 | ||
AK131594.1 | BAE20707.1 | |||
AK141724.1 | BAE24810.1 | |||
BC147444.1 | ||||
BC147468.1 |