Assay Details
Target Gene Details
Entrez Gene ID: | 66587 |
Gene Name: | Fas-activated serine/threonine kinase |
Gene Aliases: |
0610011K02Rik, 610011K02Rik |
Location: |
Chr.5:24441039-24445282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fastk | NM_023229.2 | NP_075718.2 | ||
AF168682.1 | AAF89660.1 | |||
AK002552.1 | ||||
AK003940.1 | BAB23086.3 | |||
AK038596.1 | BAC30060.1 | |||
AK052667.1 | BAC35089.1 | |||
AK139374.1 | ||||
AK151859.1 | BAE30748.1 | |||
AK152369.1 | BAE31159.1 | |||
AK159751.1 | BAE35342.1 | |||
AK167346.1 | BAE39445.1 | |||
AK177984.1 | ||||
AK179538.1 | ||||
BC013547.1 | AAH13547.1 |