Assay Details
Target Gene Details
Entrez Gene ID: | 17329 |
Gene Name: | chemokine (C-X-C motif) ligand 9 |
Gene Aliases: |
BB139920, CMK, Mig, MuMIG, Scyb9, crg-10 |
Location: |
Chr.5:92321331-92328079 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl9 | NM_008599.4 | NP_032625.2 | ||
AK042017.1 | BAC31132.1 | |||
AK042480.1 | BAC31271.1 | |||
AK156012.1 | BAE33544.1 | |||
AK156029.1 | BAE33552.1 | |||
AK156453.1 | BAE33718.1 | |||
AK157109.1 | BAE33966.1 | |||
AK157118.1 | BAE33967.1 | |||
BC003343.1 | AAH03343.1 | |||
CT010194.1 | CAJ18402.1 | |||
M34815.1 | AAA39706.1 |
More Information
Set Membership: |
Intragenic Non-exonic |