Assay Details
Target Gene Details
Entrez Gene ID: | 22385 |
Gene Name: | bromodomain adjacent to zinc finger domain, 1B |
Gene Aliases: |
C87820, WSTF, Wbscr9 |
Location: |
Chr.5:135187323-135246129 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz1b | NM_011714.2 | 6 | 1115 | NP_035844.2 |
AF084480.1 | 6 | 1074 | AAD08676.1 | |
AK017894.1 | 6 | 1116 | BAB30992.1 | |
AK036160.1 | 3 | 3261 | ||
AK137139.1 | 2 | 852 | BAE23247.1 | |
AK140172.1 | 6 | 903 | BAE24264.1 | |
AK141305.1 | 6 | 856 | BAE24643.1 | |
BC141399.1 | 6 | 1018 |