Assay Details
Target Gene Details
Entrez Gene ID: | 74610 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 8 |
Gene Aliases: |
4833412N02Rik, AA409895 |
Location: |
Chr.5:24393681-24409947 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcb8 | NM_029020.2 | NP_083296.2 | ||
AK014319.1 | BAB29270.1 | |||
AK014688.1 | ||||
AK030624.1 | BAC27052.1 | |||
AK038760.1 | ||||
AK049152.1 | BAC33571.1 | |||
AK076315.1 | BAC36297.1 | |||
AK166704.1 | BAE38958.1 | |||
AK170334.1 | ||||
AK188519.1 | ||||
AK212771.1 | ||||
BC015301.1 | AAH15301.1 |
More Information
Set Membership: |
Intragenic Non-exonic |